Product Details
- SNP ID
-
rs146268429
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:75349226 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCGCTTTTACACGGCCCCGCCTGG[C/T]CCCCGGCTCGCCCTATGTTTCGTGG
- Phenotype
-
MIM: 616701
MIM: 154580
MIM: 608844
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
COMMD4
PubMed Links
Gene Details
- Gene
- COMMD4
- Gene Name
- COMM domain containing 4
There are no transcripts associated with this gene.
- Gene
- MAN2C1
- Gene Name
- mannosidase alpha class 2C member 1
There are no transcripts associated with this gene.
- Gene
- MIR631
- Gene Name
- microRNA 631
There are no transcripts associated with this gene.
- Gene
- NEIL1
- Gene Name
- nei like DNA glycosylase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001256552.1 |
746 |
Silent Mutation |
GGC,GGT |
G193G |
NP_001243481.1 |
NM_024608.3 |
746 |
Silent Mutation |
GGC,GGT |
G107G |
NP_078884.2 |
XM_005254659.4 |
746 |
Silent Mutation |
GGC,GGT |
G107G |
XP_005254716.1 |
XM_006720677.3 |
746 |
Silent Mutation |
GGC,GGT |
G184G |
XP_006720740.1 |
XM_006720678.3 |
746 |
Silent Mutation |
GGC,GGT |
G193G |
XP_006720741.1 |
XM_006720679.3 |
746 |
Silent Mutation |
GGC,GGT |
G150G |
XP_006720742.1 |
XM_006720680.1 |
746 |
Silent Mutation |
GGC,GGT |
G107G |
XP_006720743.1 |
XM_006720681.1 |
746 |
Silent Mutation |
GGC,GGT |
G107G |
XP_006720744.1 |
XM_011522001.2 |
746 |
Silent Mutation |
GGC,GGT |
G170G |
XP_011520303.2 |
XM_011522002.1 |
746 |
Silent Mutation |
GGC,GGT |
G107G |
XP_011520304.1 |
XM_011522003.2 |
746 |
Silent Mutation |
GGC,GGT |
G107G |
XP_011520305.1 |
XM_011522004.2 |
746 |
Silent Mutation |
GGC,GGT |
G107G |
XP_011520306.1 |
XM_011522005.1 |
746 |
UTR 5 |
|
|
XP_011520307.1 |
XM_011522006.1 |
746 |
Missense Mutation |
GCC,GTC |
A12V |
XP_011520308.1 |
XM_011522007.1 |
746 |
Missense Mutation |
GCC,GTC |
A12V |
XP_011520309.1 |
XM_011522008.1 |
746 |
Missense Mutation |
GCC,GTC |
A12V |
XP_011520310.1 |
XM_017022566.1 |
746 |
Silent Mutation |
GGC,GGT |
G184G |
XP_016878055.1 |
XM_017022567.1 |
746 |
Missense Mutation |
GCC,GTC |
A12V |
XP_016878056.1 |
XM_017022568.1 |
746 |
Silent Mutation |
GGC,GGT |
G193G |
XP_016878057.1 |
View Full Product Details