Product Details

SNP ID
rs146304930
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41849880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGTGTCCCACCTGAGTAGACA[A/T]CCAGGCCGGGCATCCTTGGTCGTGC
Phenotype
MIM: 606088 MIM: 605916
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
JMJD7-PLA2G4B PubMed Links

Gene Details

Gene
JMJD7-PLA2G4B
Gene Name
JMJD7-PLA2G4B readthrough
There are no transcripts associated with this gene.

Gene
PLA2G4B
Gene Name
phospholipase A2 group IVB
There are no transcripts associated with this gene.

Gene
SPTBN5
Gene Name
spectrin beta, non-erythrocytic 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016642.3 11207 Silent Mutation GGA,GGT G3667G NP_057726.4
XM_017022299.1 11207 Silent Mutation GGA,GGT G3727G XP_016877788.1
XM_017022300.1 11207 Silent Mutation GGA,GGT G3687G XP_016877789.1
XM_017022301.1 11207 Silent Mutation GGA,GGT G3435G XP_016877790.1
XM_017022302.1 11207 Silent Mutation GGA,GGT G2786G XP_016877791.1
XM_017022303.1 11207 Intron XP_016877792.1
XM_017022304.1 11207 Intron XP_016877793.1
XM_017022305.1 11207 Intron XP_016877794.1

View Full Product Details