Product Details

SNP ID
rs146489542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40706187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTATTTTTCCCAGGCTGAGGCAG[C/T]TAAATTAGTTCCAATGGGTTTCACC
Phenotype
MIM: 179617
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAD51 PubMed Links

Gene Details

Gene
RAD51
Gene Name
RAD51 recombinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164269.1 285 Intron NP_001157741.1
NM_001164270.1 285 Missense Mutation GCT,GTT A79V NP_001157742.1
NM_002875.4 285 Missense Mutation GCT,GTT A79V NP_002866.2
NM_133487.3 285 Intron NP_597994.3
XM_006720626.3 285 Missense Mutation GCT,GTT A79V XP_006720689.1
XM_011521857.2 285 Missense Mutation GCT,GTT A79V XP_011520159.1
XM_011521858.2 285 Missense Mutation GCT,GTT A79V XP_011520160.1
XM_011521859.2 285 Missense Mutation GCT,GTT A79V XP_011520161.1
XM_011521860.2 285 Missense Mutation GCT,GTT A79V XP_011520162.1
XM_011521861.2 285 Missense Mutation GCT,GTT A79V XP_011520163.1
XM_011521862.2 285 Intron XP_011520164.1

View Full Product Details