Product Details

SNP ID
rs146667387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:99139189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTTTCAAAGGAACTTGTCCTTTA[G/T]GCAAGTTATGGGAAGTCAGCAAAAC
Phenotype
MIM: 606087
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SYNM PubMed Links

Gene Details

Gene
SYNM
Gene Name
synemin
There are no transcripts associated with this gene.

Gene
TTC23
Gene Name
tetratricopeptide repeat domain 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288615.1 Intron NP_001275544.1
NM_001288616.1 Intron NP_001275545.1
XM_011521927.2 Intron XP_011520229.1
XM_011521929.1 Intron XP_011520231.1
XM_011521930.1 Intron XP_011520232.1
XM_011521931.1 Intron XP_011520233.1
XM_011521932.1 Intron XP_011520234.1
XM_011521933.1 Intron XP_011520235.1
XM_011521934.1 Intron XP_011520236.1
XM_011521935.1 Intron XP_011520237.1
XM_011521936.1 Intron XP_011520238.1
XM_011521937.1 Intron XP_011520239.1
XM_011521939.1 Intron XP_011520241.1
XM_011521940.2 Intron XP_011520242.1
XM_011521941.2 Intron XP_011520243.1
XM_011521943.2 Intron XP_011520245.1
XM_011521944.2 Intron XP_011520246.1
XM_011521945.2 Intron XP_011520247.1
XM_017022509.1 Intron XP_016877998.1
XM_017022510.1 Intron XP_016877999.1
XM_017022511.1 Intron XP_016878000.1
XM_017022512.1 Intron XP_016878001.1
XM_017022513.1 Intron XP_016878002.1
XM_017022514.1 Intron XP_016878003.1
XM_017022515.1 Intron XP_016878004.1
XM_017022516.1 Intron XP_016878005.1
XM_017022517.1 Intron XP_016878006.1
XM_017022518.1 Intron XP_016878007.1

View Full Product Details