Product Details

SNP ID
rs146679421
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40807429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCGTACAGGTCCATCTGTAAGAG[A/C]TCCTTGGTCACTGCCATGGTTCCGG
Phenotype
MIM: 616844 MIM: 613256
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC17 PubMed Links

Gene Details

Gene
DNAJC17
Gene Name
DnaJ heat shock protein family (Hsp40) member C17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018163.2 354 Missense Mutation NP_060633.1
XM_017022401.1 354 UTR 5 XP_016877890.1
XM_017022402.1 354 Intron XP_016877891.1
Gene
PPP1R14D
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14D
There are no transcripts associated with this gene.

Gene
ZFYVE19
Gene Name
zinc finger FYVE-type containing 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077268.1 354 UTR 5 NP_001070736.1
NM_001258420.1 354 UTR 5 NP_001245349.1
NM_001258421.1 354 UTR 5 NP_001245350.1
NM_032850.4 354 Intron NP_116239.3
XM_017022683.1 354 Intron XP_016878172.1
XM_017022684.1 354 UTR 5 XP_016878173.1

View Full Product Details