Product Details

SNP ID
rs147009618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40695039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGGGAAGAGGGCAGTCTGTAA[A/G]CTCGCGCAGGATCAAGCTCTCGAGC
Phenotype
MIM: 179617
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAD51 PubMed Links

Gene Details

Gene
RAD51
Gene Name
RAD51 recombinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164269.1 Intron NP_001157741.1
NM_001164270.1 Intron NP_001157742.1
NM_002875.4 Intron NP_002866.2
NM_133487.3 Intron NP_597994.3
XM_006720626.3 Intron XP_006720689.1
XM_011521857.2 Intron XP_011520159.1
XM_011521858.2 Intron XP_011520160.1
XM_011521859.2 Intron XP_011520161.1
XM_011521860.2 Intron XP_011520162.1
XM_011521861.2 Intron XP_011520163.1
XM_011521862.2 Intron XP_011520164.1
Gene
RAD51-AS1
Gene Name
RAD51 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details