Product Details
- SNP ID
-
rs147579391
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:59138302 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTGCACTGTGGCACCTGAGGCTTG[G/T]GCTTGGGCTGGGGCTTGGGTCTGCC
- Phenotype
-
MIM: 601479
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
MYO1E
PubMed Links
Gene Details
- Gene
- MYO1E
- Gene Name
- myosin IE
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004998.3 |
3518 |
Missense Mutation |
CAC,CCC |
H1049P |
NP_004989.2 |
View Full Product Details