Product Details

SNP ID
rs147604059
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:26544041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTGCCCCAAGATTCGATGTTAC[A/T]CTAATGGCTCACACTGACAGCCAGA
Phenotype
MIM: 137192
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GABRB3 PubMed Links

Gene Details

Gene
GABRB3
Gene Name
gamma-aminobutyric acid type A receptor beta3 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000814.5 5384 UTR 3 NP_000805.1
NM_001191320.1 5384 UTR 3 NP_001178249.1
NM_001191321.2 5384 UTR 3 NP_001178250.1
NM_001278631.1 5384 UTR 3 NP_001265560.1
NM_021912.4 5384 UTR 3 NP_068712.1
XM_011521428.2 5384 UTR 3 XP_011519730.1

View Full Product Details