Product Details

SNP ID
rs147692284
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41900723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGCTGCTGGGCAGCTCGTAGCC[A/G]TCGAGCTTGATCTTGATGAGGTGCT
Phenotype
MIM: 605892 MIM: 605916
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EHD4 PubMed Links

Gene Details

Gene
EHD4
Gene Name
EH domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139265.3 1611 Silent Mutation GAC,GAT D516D NP_644670.1
XM_017022100.1 1611 Silent Mutation GAC,GAT D428D XP_016877589.1
Gene
LOC105370792
Gene Name
uncharacterized LOC105370792
There are no transcripts associated with this gene.

Gene
SPTBN5
Gene Name
spectrin beta, non-erythrocytic 5
There are no transcripts associated with this gene.

View Full Product Details