Product Details

SNP ID
rs148042984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42415505 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAAAAGAACACATACTCAGTCTC[A/T]CACACAATTTCTGGGGGCTCACAGA
Phenotype
MIM: 114240
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CAPN3 PubMed Links

Gene Details

Gene
CAPN3
Gene Name
calpain 3
There are no transcripts associated with this gene.

Gene
ZNF106
Gene Name
zinc finger protein 106
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284306.1 7240 UTR 3 NP_001271235.1
NM_001284307.1 7240 UTR 3 NP_001271236.1
NM_022473.2 7240 UTR 3 NP_071918.1
XM_005254591.2 7240 UTR 3 XP_005254648.1
XM_005254592.2 7240 Intron XP_005254649.1
XM_017022483.1 7240 Intron XP_016877972.1
XM_017022484.1 7240 UTR 3 XP_016877973.1
XM_017022485.1 7240 UTR 3 XP_016877974.1

View Full Product Details