Product Details

SNP ID
rs148575688
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:32643788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGAATATCCAGCTCACCAGGCC[A/T]TGAATCTTGTGGGCCCCCAGAGCAT
Phenotype
MIM: 610589 MIM: 173120
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP11A PubMed Links

Gene Details

Gene
ARHGAP11A
Gene Name
Rho GTPase activating protein 11A
There are no transcripts associated with this gene.

Gene
SCG5
Gene Name
secretogranin V
There are no transcripts associated with this gene.

View Full Product Details