Product Details

SNP ID
rs148849981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:59618454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGAGGTCTATCAACTGTTCAGG[A/G]GTCACCCGAGGGGACCAAGAGGCAG
Phenotype
MIM: 606836
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GCNT3 PubMed Links

Gene Details

Gene
GCNT3
Gene Name
glucosaminyl (N-acetyl) transferase 3, mucin type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004751.2 665 Silent Mutation GGA,GGG G72G NP_004742.1

View Full Product Details