Product Details

SNP ID
rs149022137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:67066163 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCGCCCTCCCCAGCCATGTCGTC[C/T]ATCCTGCCTTTCACTCCCCCGATCG
Phenotype
MIM: 603109
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC102723493 PubMed Links

Gene Details

Gene
LOC102723493
Gene Name
uncharacterized LOC102723493
There are no transcripts associated with this gene.

Gene
SMAD3
Gene Name
SMAD family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145102.1 307 Intron NP_001138574.1
NM_001145103.1 307 Intron NP_001138575.1
NM_001145104.1 307 Intron NP_001138576.1
NM_005902.3 307 Silent Mutation TCC,TCT S3S NP_005893.1
XM_011521559.2 307 Silent Mutation TCC,TCT S3S XP_011519861.1
XM_011521560.1 307 Intron XP_011519862.1

View Full Product Details