Product Details

SNP ID
rs149103241
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:39582426 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCTGAGGCAGATGAAGAAGACC[C/G]GGGGCACGCTGCTGGCCCTGGAGCG
Phenotype
MIM: 188060
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
THBS1 PubMed Links

Gene Details

Gene
THBS1
Gene Name
thrombospondin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003246.3 476 Missense Mutation CGG,GGG R101G NP_003237.2
XM_011521971.1 476 Missense Mutation CGG,GGG R101G XP_011520273.1

View Full Product Details