Product Details

SNP ID
rs150257540
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:44551510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAACAGAAGAAAAGGCAAGAAGA[A/T]ATTAAAAAGAGGGTAAATTCTGTTT
Phenotype
MIM: 603910
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
EIF3J PubMed Links

Gene Details

Gene
EIF3J
Gene Name
eukaryotic translation initiation factor 3 subunit J
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284335.1 409 Missense Mutation GAA,GAT E94D NP_001271264.1
NM_001284336.1 409 Intron NP_001271265.1
NM_003758.3 409 Missense Mutation GAA,GAT E94D NP_003749.2

View Full Product Details