Product Details

SNP ID
rs150369977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:79845439 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGCATGAAGATGAGATCAAGT[C/T]CCCCTGCGGAAAAGAGGAACAAATT
Phenotype
MIM: 604197
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTHFS PubMed Links

Gene Details

Gene
MTHFS
Gene Name
5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199758.1 390 Missense Mutation GAA,GGA E71G NP_001186687.1
NM_006441.3 390 Missense Mutation GAA,GGA E128G NP_006432.1
Gene
ST20-MTHFS
Gene Name
ST20-MTHFS readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199760.1 390 Missense Mutation GAA,GGA E104G NP_001186689.1

View Full Product Details