Product Details

SNP ID
rs150453767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:63325583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCATTCTCTTTGTCCCTCTAGCT[C/G]TCTCTCCCTTCTCATCCCCACCAAG
Phenotype
MIM: 603263
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CA12 PubMed Links

Gene Details

Gene
CA12
Gene Name
carbonic anhydrase 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001218.4 2124 UTR 3 NP_001209.1
NM_001293642.1 2124 UTR 3 NP_001280571.1
NM_206925.2 2124 UTR 3 NP_996808.1

View Full Product Details