Product Details

SNP ID
rs150929841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64848608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTTACAGGGTGTGAAGGAAGCC[A/G]GTGAGAAGCCTCGGGGAGCACAGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLEKHO2 PubMed Links

Gene Details

Gene
PLEKHO2
Gene Name
pleckstrin homology domain containing O2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195059.1 162 Intron NP_001181988.1
NM_025201.4 162 Missense Mutation AGT,GGT S10G NP_079477.2

View Full Product Details