Product Details

SNP ID
rs151337601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73740004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTGACACTGTCTGGGTGCTGCT[C/G]TTCCTCGTGACCCTTCGAGTCTGTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C15orf59 PubMed Links

Gene Details

Gene
C15orf59
Gene Name
chromosome 15 open reading frame 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039614.2 1023 Missense Mutation AAC,AAG N265K NP_001034703.1
NM_001303254.1 1023 Missense Mutation AAC,AAG N237K NP_001290183.1
XM_005254369.4 1023 Missense Mutation AAC,AAG N265K XP_005254426.1

View Full Product Details