Product Details
- SNP ID
-
rs76909004
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:30959752 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGGGGTCCCTTCATTCTTCTTCTT[C/T]TTTTTTTTTTTTTTACTTTGTACAC
- Phenotype
-
MIM: 610930
MIM: 611052
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ORAI3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs140020755] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ORAI3
- Gene Name
- ORAI calcium release-activated calcium modulator 3
There are no transcripts associated with this gene.
- Gene
- SETD1A
- Gene Name
- SET domain containing 1A
View Full Product Details