Product Details

SNP ID
rs138136838
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:46709638 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGGCAAATTATTCAGCCACTC[A/G]TGTTTCAGGCACTGTGTGGCACTCA
Phenotype
MIM: 612147
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYLK3 PubMed Links

Gene Details

Gene
MYLK3
Gene Name
myosin light chain kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308301.1 1598 Silent Mutation CAC,CAT H426H NP_001295230.1
NM_182493.2 1598 Silent Mutation CAC,CAT H767H NP_872299.2
XM_006721334.3 1598 Silent Mutation CAC,CAT H426H XP_006721397.1

View Full Product Details