Product Details

SNP ID
rs138192595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3356454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCAGCTCCCATTGTGTGGGCAC[C/T]GGAGGGTGGAGGGATTCCTCTGCGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2C1 PubMed Links

Gene Details

Gene
OR2C1
Gene Name
olfactory receptor family 2 subfamily C member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012368.2 566 Missense Mutation CGG,TGG R172W NP_036500.2

View Full Product Details