Product Details

SNP ID
rs138284829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:22347486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCTCTGGCTGGGCTCTTTGAAAG[C/G]AACATACAGAGAGTCTGGCACCAGC
Phenotype
MIM: 117340
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDR2 PubMed Links

Gene Details

Gene
CDR2
Gene Name
cerebellar degeneration related protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001802.1 1152 Missense Mutation CCT,GCT P282A NP_001793.1

View Full Product Details