Product Details
- SNP ID
-
rs138416464
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:30651943 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCGGGTCCTGGCCGTGGTGCTAGA[A/T]GATGTCATGGCTGTTCACATGGTGA
- Phenotype
-
MIM: 608601
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FBRS
PubMed Links
Gene Details
- Gene
- FBRS
- Gene Name
- fibrosin
There are no transcripts associated with this gene.
- Gene
- PRR14
- Gene Name
- proline rich 14
View Full Product Details