Product Details

SNP ID
rs138417092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:31084678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCCCTAGGCCCCAGGAGGGTCAC[A/G]GGCACTGTCTGGAGGGAGCTGATGC
Phenotype
MIM: 610561 MIM: 608547
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRSS53 PubMed Links

Gene Details

Gene
PRSS53
Gene Name
protease, serine 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039503.2 2531 Silent Mutation CCC,CCT P435P NP_001034592.1
XM_011545816.1 2531 Silent Mutation CCC,CCT P482P XP_011544118.1
XM_011545817.1 2531 Silent Mutation CCC,CCT P482P XP_011544119.1
XM_011545818.2 2531 Silent Mutation CCC,CCT P482P XP_011544120.1
XM_011545819.1 2531 Silent Mutation CCC,CCT P435P XP_011544121.1
XM_011545820.1 2531 Silent Mutation CCC,CCT P435P XP_011544122.1
Gene
VKORC1
Gene Name
vitamin K epoxide reductase complex subunit 1
There are no transcripts associated with this gene.

Gene
ZNF646
Gene Name
zinc finger protein 646
There are no transcripts associated with this gene.

View Full Product Details