Product Details

SNP ID
rs138418408
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:24255627 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTTTAAGAAGACTCGGTCTTTC[G/T]GGTCTTCTTTTTCCCCAATGGGCTC
Phenotype
MIM: 606403
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CACNG3 PubMed Links

Gene Details

Gene
CACNG3
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006539.3 75 UTR 5 NP_006530.1

View Full Product Details