Product Details
- SNP ID
-
rs138557834
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:67908795 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTCCTTCCCGAGCCACCCAAGGAT[A/G]TCCAGCTGGATCTGGTCAAGAAGGT
- Phenotype
-
MIM: 177015
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PSKH1
PubMed Links
Gene Details
- Gene
- PSKH1
- Gene Name
- protein serine kinase H1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006742.2 |
216 |
Missense Mutation |
ATC,GTC |
I16V |
NP_006733.1 |
View Full Product Details