Product Details

SNP ID
rs138811771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4611182 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGGTTCGACTAGGTGGCTCCT[A/G]GGTCCCCTCCCTTGCAACAGTGACC
Phenotype
MIM: 607559
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MGRN1 PubMed Links

Gene Details

Gene
MGRN1
Gene Name
mahogunin ring finger 1
There are no transcripts associated with this gene.

Gene
UBALD1
Gene Name
UBA like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145253.2 Intron NP_660296.1
XM_017022937.1 Intron XP_016878426.1

View Full Product Details