Product Details

SNP ID
rs138910759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:72013486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCGTGGGCCCACACTTGCAGC[A/G]GCTCATTTAGAATGTGACTCAGACT
Phenotype
MIM: 126064
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHODH PubMed Links

Gene Details

Gene
DHODH
Gene Name
dihydroorotate dehydrogenase (quinone)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001361.4 Intron NP_001352.2
XM_005255827.3 Intron XP_005255884.1
XM_005255829.3 Intron XP_005255886.1
XM_017022990.1 Intron XP_016878479.1

View Full Product Details