Product Details

SNP ID
rs139036864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68245496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCTCCTGTTCCTCTTGCTGCTG[C/T]TAATGCTGCTCGCGGACCCAGCGCT
Phenotype
MIM: 612960 MIM: 609362
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESRP2 PubMed Links

Gene Details

Gene
ESRP2
Gene Name
epithelial splicing regulatory protein 2
There are no transcripts associated with this gene.

Gene
PLA2G15
Gene Name
phospholipase A2 group XV
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012320.3 126 Silent Mutation CTA,TTA L24L NP_036452.1
XM_005255886.3 126 Silent Mutation CTA,TTA L24L XP_005255943.1
XM_011522979.2 126 Silent Mutation CTA,TTA L24L XP_011521281.1
XM_011522980.2 126 UTR 5 XP_011521282.2

View Full Product Details