Product Details

SNP ID
rs139471830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2818852 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACATTGCCTTGGTGAAGCTGTCT[C/G]CACCTGTCACCTACACTAAACACAT
Phenotype
MIM: 608159
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PRSS21 PubMed Links

Gene Details

Gene
PRSS21
Gene Name
protease, serine 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270452.1 459 Missense Mutation CCA,GCA P143A NP_001257381.1
NM_006799.3 459 Missense Mutation CCA,GCA P145A NP_006790.1
NM_144956.2 459 Missense Mutation CCA,GCA P143A NP_659205.1
NM_144957.2 459 Missense Mutation CCA,GCA P145A NP_659206.1
XM_017022875.1 459 Missense Mutation CCA,GCA P143A XP_016878364.1
XM_017022876.1 459 Missense Mutation CCA,GCA P145A XP_016878365.1
XM_017022877.1 459 Missense Mutation CCA,GCA P143A XP_016878366.1
XM_017022878.1 459 Missense Mutation CCA,GCA P145A XP_016878367.1

View Full Product Details