Product Details

SNP ID
rs139649973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30555844 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCGCCAGTGTGACTGTAGACGT[A/G]CTCCACCAGTGTGGAGCGCCAGGCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF764 PubMed Links

Gene Details

Gene
ZNF764
Gene Name
zinc finger protein 764
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172679.1 716 Missense Mutation CAC,TAC H192Y NP_001166150.1
NM_033410.3 716 Missense Mutation CAC,TAC H193Y NP_219363.2

View Full Product Details