Product Details

SNP ID
rs139688166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30958869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGTACCGCTATGATGGAGTCCA[C/T]TTCAGTGTCAACGTGAGTGCCCGGG
Phenotype
MIM: 610930 MIM: 611052
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ORAI3 PubMed Links

Gene Details

Gene
ORAI3
Gene Name
ORAI calcium release-activated calcium modulator 3
There are no transcripts associated with this gene.

Gene
SETD1A
Gene Name
SET domain containing 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014712.2 229 Silent Mutation CAC,CAT H46H NP_055527.1
XM_005255723.1 229 Silent Mutation CAC,CAT H46H XP_005255780.1
XM_006721106.3 229 Silent Mutation CAC,CAT H46H XP_006721169.1
XM_017023909.1 229 Silent Mutation CAC,CAT H46H XP_016879398.1

View Full Product Details