Product Details

SNP ID
rs139719193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75628635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGCCCATGCCCCAGCCAGCTGT[G/T]GGGGGCAGCCCATATTCCAGGGCAG
Phenotype
MIM: 604230 MIM: 601421
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ADAT1 PubMed Links

Gene Details

Gene
ADAT1
Gene Name
adenosine deaminase, tRNA specific 1
There are no transcripts associated with this gene.

Gene
KARS
Gene Name
lysyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130089.1 1769 Silent Mutation CCA,CCC P571P NP_001123561.1
NM_005548.2 1769 Intron NP_005539.1
XM_017023217.1 1769 Silent Mutation CCA,CCC P387P XP_016878706.1

View Full Product Details