Product Details

SNP ID
rs139866691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:68738336 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCAGGAGCCGGAGCCCTGCCAC[A/C]CTGGCTTTGACGCCGAGAGCTACAC
Phenotype
MIM: 192090
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDH1 PubMed Links

Gene Details

Gene
CDH1
Gene Name
cadherin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317184.1 214 Missense Mutation ACT,CCT T30P NP_001304113.1
NM_001317185.1 214 UTR 5 NP_001304114.1
NM_001317186.1 214 UTR 5 NP_001304115.1
NM_004360.4 214 Missense Mutation ACT,CCT T30P NP_004351.1
XM_011523488.1 214 Intron XP_011521790.1
XM_011523489.1 214 Intron XP_011521791.1

View Full Product Details