Product Details

SNP ID
rs140080780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1980021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAGGGGTGCCGCAGCAGCACGT[A/C]CAGGCTCTCCTGGGCCTGCGCCTGA
Phenotype
MIM: 600924 MIM: 611256 MIM: 603927 MIM: 605915
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GFER PubMed Links

Gene Details

Gene
GFER
Gene Name
growth factor, augmenter of liver regeneration
There are no transcripts associated with this gene.

Gene
NOXO1
Gene Name
NADPH oxidase organizer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267721.1 944 Missense Mutation GAC,TAC D192Y NP_001254650.1
NM_144603.3 944 Missense Mutation GAC,TAC D187Y NP_653204.1
NM_172167.2 944 Missense Mutation GAC,TAC D188Y NP_751907.1
NM_172168.2 944 Missense Mutation GAC,TAC D193Y NP_751908.1
XM_005255099.4 944 Missense Mutation GAC,TAC D193Y XP_005255156.1
XM_017022927.1 944 Missense Mutation GAC,TAC D193Y XP_016878416.1
XM_017022928.1 944 Missense Mutation GAC,TAC D193Y XP_016878417.1
Gene
SYNGR3
Gene Name
synaptogyrin 3
There are no transcripts associated with this gene.

Gene
TBL3
Gene Name
transducin beta like 3
There are no transcripts associated with this gene.

View Full Product Details