Product Details

SNP ID
rs140112104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71850466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGGCTACCTGCTGGGTATACTTT[G/T]CATGAAACCCCTCCAGCAGGTGCCA
Phenotype
MIM: 614301
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ATXN1L PubMed Links

Gene Details

Gene
ATXN1L
Gene Name
ataxin 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137675.3 1019 Missense Mutation TTG,TTT L242F NP_001131147.1
Gene
ZNF821
Gene Name
zinc finger protein 821
There are no transcripts associated with this gene.

View Full Product Details