Product Details

SNP ID
rs140173167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67003181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACTGAAAGTGGCCAAGGTGAGT[G/T]CCTCTCCTTCCCCAGGGCTCAGCAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CES4A PubMed Links

Gene Details

Gene
CES4A
Gene Name
carboxylesterase 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190201.1 Intron NP_001177130.1
NM_001190202.1 Intron NP_001177131.1
NM_001318506.1 Intron NP_001305435.1
NM_173815.6 Intron NP_776176.5
XM_005255896.4 Intron XP_005255953.1
XM_011523021.2 Intron XP_011521323.1
XM_011523023.2 Intron XP_011521325.1
XM_011523024.2 Intron XP_011521326.1
XM_011523025.2 Intron XP_011521327.1
XM_011523026.2 Intron XP_011521328.1
XM_011523027.2 Intron XP_011521329.1
XM_011523028.2 Intron XP_011521330.1
XM_011523029.2 Intron XP_011521331.1
XM_011523030.2 Intron XP_011521332.1
XM_017023160.1 Intron XP_016878649.1
XM_017023161.1 Intron XP_016878650.1
XM_017023162.1 Intron XP_016878651.1
XM_017023163.1 Intron XP_016878652.1
XM_017023164.1 Intron XP_016878653.1
XM_017023165.1 Intron XP_016878654.1
XM_017023166.1 Intron XP_016878655.1
XM_017023167.1 Intron XP_016878656.1
XM_017023168.1 Intron XP_016878657.1

View Full Product Details