Product Details

SNP ID
rs140275778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67934827 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGGCTCTGTGGGTGAGCTCAGT[A/G]TCCGCAGCAGCTTGGCGCCAGTCTT
Phenotype
MIM: 118888 MIM: 606967 MIM: 177015 MIM: 176847 MIM: 604119
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTRL PubMed Links

Gene Details

Gene
CTRL
Gene Name
chymotrypsin like
There are no transcripts associated with this gene.

Gene
LCAT
Gene Name
lecithin-cholesterol acyltransferase
There are no transcripts associated with this gene.

Gene
PSKH1
Gene Name
protein serine kinase H1
There are no transcripts associated with this gene.

Gene
PSMB10
Gene Name
proteasome subunit beta 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002801.3 808 Missense Mutation ACA,ATA T227I NP_002792.1
Gene
SLC12A4
Gene Name
solute carrier family 12 member 4
There are no transcripts associated with this gene.

View Full Product Details