Product Details

SNP ID
rs140339652
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:166692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACTGGTGTGGCCGTGGTGCTGA[C/G]CGAAAAATACCGCTGAGCCCTGTGC
Phenotype
MIM: 141800 MIM: 141850 MIM: 609639
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HBA1 PubMed Links

Gene Details

Gene
HBA1
Gene Name
hemoglobin subunit alpha 1
There are no transcripts associated with this gene.

Gene
HBA2
Gene Name
hemoglobin subunit alpha 2
There are no transcripts associated with this gene.

Gene
HBM
Gene Name
hemoglobin subunit mu
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003938.3 434 Missense Mutation ACC,AGC T137S NP_001003938.1

View Full Product Details