Product Details

SNP ID
rs140442350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:676431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCCCTGGACCCGGCCAAGCTGGA[A/C]ATGCTGGTGGCCCTGGCTCAGAGCA
Phenotype
MIM: 603264 MIM: 613889 MIM: 607207
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
JMJD8 PubMed Links

Gene Details

Gene
JMJD8
Gene Name
jumonji domain containing 8
There are no transcripts associated with this gene.

Gene
LOC105371184
Gene Name
uncharacterized LOC105371184
There are no transcripts associated with this gene.

Gene
RHBDL1
Gene Name
rhomboid like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278720.1 360 Silent Mutation GAA,GAC E45D NP_001265649.1
NM_001278721.1 360 Silent Mutation GAA,GAC E45D NP_001265650.1
NM_001318733.1 360 Silent Mutation GAA,GAC E110D NP_001305662.1
XM_017023849.1 360 Missense Mutation GAA,GAC E110D XP_016879338.1
XM_017023850.1 360 Missense Mutation GAA,GAC E110D XP_016879339.1
Gene
RHOT2
Gene Name
ras homolog family member T2
There are no transcripts associated with this gene.

Gene
STUB1
Gene Name
STIP1 homology and U-box containing protein 1
There are no transcripts associated with this gene.

Gene
WDR24
Gene Name
WD repeat domain 24
There are no transcripts associated with this gene.

Gene
WDR90
Gene Name
WD repeat domain 90
There are no transcripts associated with this gene.

View Full Product Details