Product Details

SNP ID
rs140461033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:66938061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGGACTCAGCCAGTCCCATCC[A/G]GACCACACACACGGGGCAGGTGCTG
Phenotype
MIM: 605278 MIM: 614778
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CES2 PubMed Links

Gene Details

Gene
CES2
Gene Name
carboxylesterase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003869.5 1293 Missense Mutation CAG,CGG Q98R NP_003860.2
NM_198061.2 1293 Missense Mutation CAG,CGG Q98R NP_932327.1
XM_011523421.2 1293 UTR 5 XP_011521723.1
XM_017023818.1 1293 UTR 5 XP_016879307.1
Gene
FAM96B
Gene Name
family with sequence similarity 96 member B
There are no transcripts associated with this gene.

View Full Product Details