Product Details

SNP ID
rs140779931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28981478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTCTGTCTCCCAGGTGACACC[A/G]GGTCTAGGAGTGGTGGCCGTTCTGC
Phenotype
MIM: 602354 MIM: 612583
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAT PubMed Links

Gene Details

Gene
LAT
Gene Name
linker for activation of T-cells
There are no transcripts associated with this gene.

Gene
SPNS1
Gene Name
sphingolipid transporter 1 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142448.1 1049 Silent Mutation CCA,CCG P224P NP_001135920.1
NM_001142449.1 1049 Silent Mutation CCA,CCG P202P NP_001135921.1
NM_001142450.1 1049 Silent Mutation CCA,CCG P151P NP_001135922.1
NM_001142451.1 1049 Silent Mutation CCA,CCG P224P NP_001135923.1
NM_032038.2 1049 Silent Mutation CCA,CCG P224P NP_114427.1
XM_006721096.3 1049 Silent Mutation CCA,CCG P269P XP_006721159.1
XM_017023757.1 1049 Silent Mutation CCA,CCG P274P XP_016879246.1
XM_017023758.1 1049 Silent Mutation CCA,CCG P274P XP_016879247.1
XM_017023759.1 1049 Silent Mutation CCA,CCG P269P XP_016879248.1

View Full Product Details