Product Details

SNP ID
rs140916935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29791820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCTCTGCTCTTAACCATTATGCC[C/T]AAGTACATGTTATCCCTGCTGGCTG
Phenotype
MIM: 603213
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF22 PubMed Links

Gene Details

Gene
KIF22
Gene Name
kinesin family member 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256269.1 Intron NP_001243198.1
NM_001256270.1 Intron NP_001243199.1
NM_007317.2 Intron NP_015556.1

View Full Product Details