Product Details

SNP ID
rs141079252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29779518 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCCCTCCAGTTCAGGCCAGGTC[C/T]TCCTCCTATAGTGGAGAGTATGGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZG16 PubMed Links

Gene Details

Gene
ZG16
Gene Name
zymogen granule protein 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152338.3 222 Silent Mutation TCC,TCT S23S NP_689551.2
XM_011545921.2 222 Silent Mutation TCC,TCT S23S XP_011544223.1

View Full Product Details