Product Details

SNP ID
rs141175891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3222559 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTAGAAAAATTTTCACAGGAATAT[A/C]ATACTCTAATAAATAGAAAGGCATG
Phenotype
MIM: 603231
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR1F2P PubMed Links

Gene Details

Gene
OR1F2P
Gene Name
olfactory receptor family 1 subfamily F member 2 pseudogene
There are no transcripts associated with this gene.

Gene
ZNF200
Gene Name
zinc finger protein 200
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145446.1 2945 UTR 3 NP_001138918.1
NM_001145447.1 2945 UTR 3 NP_001138919.1
NM_001145448.1 2945 UTR 3 NP_001138920.1
NM_003454.3 2945 UTR 3 NP_003445.2
NM_198087.2 2945 UTR 3 NP_932353.1
NM_198088.2 2945 UTR 3 NP_932354.1
XM_005255556.1 2945 UTR 3 XP_005255613.1
XM_006720940.2 2945 UTR 3 XP_006721003.1
XM_006720941.2 2945 UTR 3 XP_006721004.1
XM_017023647.1 2945 UTR 3 XP_016879136.1
XM_017023648.1 2945 Intron XP_016879137.1

View Full Product Details