Product Details

SNP ID
rs141226927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:78022719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACATCCAGATTAAAAGCCAGGAA[C/G]CACAGCAAACGTCGAGTGAGAGGTA
Phenotype
MIM: 613588
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLEC3A PubMed Links

Gene Details

Gene
CLEC3A
Gene Name
C-type lectin domain family 3 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244755.1 174 Intron NP_001231684.1
NM_005752.4 174 Missense Mutation AAC,AAG N40K NP_005743.4

View Full Product Details