Product Details

SNP ID
rs141460735
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1790902 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGGCTGAGGTAGCGCAGCCGCC[C/T]CAGTGGTGCCAAGAGGCTGTTGGGC
Phenotype
MIM: 601489 MIM: 610779 MIM: 611659
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGFALS PubMed Links

Gene Details

Gene
IGFALS
Gene Name
insulin like growth factor binding protein acid labile subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146006.1 1711 Missense Mutation AGG,GGG R544G NP_001139478.1
NM_004970.2 1711 Missense Mutation AGG,GGG R506G NP_004961.1
Gene
NUBP2
Gene Name
nucleotide binding protein 2
There are no transcripts associated with this gene.

Gene
SPSB3
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
There are no transcripts associated with this gene.

View Full Product Details