Product Details

SNP ID
rs141629916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:23392458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGATCGCATCACAGTAGGTCTGCA[C/T]TGTGGCTCTGGCGATCGAGCCCAGC
Phenotype
MIM: 606978
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COG7 PubMed Links

Gene Details

Gene
COG7
Gene Name
component of oligomeric golgi complex 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153603.3 2265 Missense Mutation ATG,GTG M690V NP_705831.1
XM_017023870.1 2265 Missense Mutation ATG,GTG M625V XP_016879359.1

View Full Product Details