Product Details

SNP ID
rs141653862
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19713657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATACAGGCATCTCCGGCTACCC[A/C]AATCTATTTGATGTATAAATTGTGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IQCK PubMed Links

Gene Details

Gene
IQCK
Gene Name
IQ motif containing K
There are no transcripts associated with this gene.

Gene
KNOP1
Gene Name
lysine rich nucleolar protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012991.2 Intron NP_001013009.2
XM_005255317.4 Intron XP_005255374.1
XM_006721046.3 Intron XP_006721109.1
XM_011545855.1 Intron XP_011544157.1
XM_017023231.1 Intron XP_016878720.1
XM_017023232.1 Intron XP_016878721.1

View Full Product Details